Product Details

SNP ID
rs200464363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49128859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGTGCTTGCAGCTGCTCCTGGA[A/C]CACCTGGAGTGCCTGGTGTCCAGGC
Phenotype
MIM: 603144
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
There are no transcripts associated with this gene.

Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 703 Missense Mutation GAA,GAC E118D NP_003651.1
XM_017027407.1 703 Missense Mutation GAA,GAC E118D XP_016882896.1

View Full Product Details