Product Details

SNP ID
rs199926479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14827355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTATTCCCAGAGCCCTCTTTATG[C/T]CTTTATTCCTCAGACTATAGATAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7A5 PubMed Links

Gene Details

Gene
OR7A5
Gene Name
olfactory receptor family 7 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017506.1 1024 Missense Mutation GAC,GGC D296G NP_059976.1
XM_011527912.2 1024 Missense Mutation GAC,GGC D296G XP_011526214.1
XM_011527913.2 1024 Missense Mutation GAC,GGC D296G XP_011526215.1
XM_017026583.1 1024 Missense Mutation GAC,GGC D340G XP_016882072.1
Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198944.1 1024 Intron NP_945182.1
XM_005259853.3 1024 Intron XP_005259910.1
XM_011527915.2 1024 Intron XP_011526217.1

View Full Product Details