Product Details

SNP ID
rs199508051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46124313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGAGGAAGACTGTTATCCAGC[A/C]GACAAGAGCTAAGGGAGAAAGGAAA
Phenotype
MIM: 610545 MIM: 610546
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGFL2 PubMed Links

Gene Details

Gene
IGFL2
Gene Name
IGF like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002915.2 2486 Intron NP_001002915.2
NM_001135113.1 2486 Intron NP_001128585.1
XM_011526501.1 2486 Intron XP_011524803.1
XM_017026335.1 2486 Intron XP_016881824.1
Gene
IGFL3
Gene Name
IGF like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207393.1 2486 Missense Mutation GGC,TGC G12C NP_997276.1
XM_011526953.2 2486 Missense Mutation GGC,TGC G12C XP_011525255.1
XM_017026796.1 2486 Missense Mutation GGC,TGC G12C XP_016882285.1
XM_017026797.1 2486 Missense Mutation GGC,TGC G12C XP_016882286.1

View Full Product Details