Product Details

SNP ID
rs199895704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45075734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTGAGCCCTGGCCGCGGCTGG[A/G]GCCTCTGAAGAGCTGACAGCCCAGC
Phenotype
MIM: 607419 MIM: 613226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLASRP PubMed Links

Gene Details

Gene
CLASRP
Gene Name
CLK4 associating serine/arginine rich protein
There are no transcripts associated with this gene.

Gene
GEMIN7
Gene Name
gem nuclear organelle associated protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007269.1 Intron NP_001007270.1
NM_001007270.1 Intron NP_001007271.1
NM_001319054.1 Intron NP_001305983.1
NM_001319055.1 Intron NP_001305984.1
NM_024707.2 Intron NP_078983.1
XM_005259262.3 Intron XP_005259319.1
XM_005259263.4 Intron XP_005259320.1
XM_017027312.1 Intron XP_016882801.1
Gene
LOC105372419
Gene Name
uncharacterized LOC105372419
There are no transcripts associated with this gene.

Gene
ZNF296
Gene Name
zinc finger protein 296
There are no transcripts associated with this gene.

View Full Product Details