Product Details

SNP ID
rs200035337
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57779488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGAATGTGGGAAATCCTTTAGC[G/T]TAAGGTCCAACCTCATTCACCATCA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF586 PubMed Links

Gene Details

Gene
ZNF586
Gene Name
zinc finger protein 586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077426.2 1092 UTR 3 NP_001070894.1
NM_001204814.1 1092 Missense Mutation GTA,TTA V258L NP_001191743.1
NM_017652.3 1092 Missense Mutation GTA,TTA V301L NP_060122.2

View Full Product Details