Product Details
- SNP ID
-
rs199617146
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56190073 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGGGCGAACCGCTTTTGGCAGAC[A/G]TCACACATGTAGGGCCTCTCGCCAG
- Phenotype
-
MIM: 611178
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GALP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16987048] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GALP
- Gene Name
- galanin like peptide
There are no transcripts associated with this gene.
- Gene
- ZSCAN5B
- Gene Name
- zinc finger and SCAN domain containing 5B
View Full Product Details