Product Details

SNP ID
rs200318517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48644440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCCCCTTACCTTCTCTCCTCCA[A/G]TGCTGAGCCTTAATGGGGGCAGAAA
Phenotype
MIM: 604644 MIM: 124097
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA11 PubMed Links

Gene Details

Gene
CA11
Gene Name
carbonic anhydrase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001217.4 952 Missense Mutation ACT,ATT T91I NP_001208.2
Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
There are no transcripts associated with this gene.

Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details