Product Details

SNP ID
rs200386941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40742489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCTGGCCACAGAGGCCAGCCC[A/G]TACCCCACCCACGGGCACCACGTAC
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 1576 Missense Mutation CGG,TGG R334W NP_940878.3
XM_005258776.4 1576 UTR 3 XP_005258833.1
XM_005258778.4 1576 Missense Mutation CGG,TGG R162W XP_005258835.1
XM_006723152.2 1576 Intron XP_006723215.1
XM_006723153.2 1576 Intron XP_006723216.1
XM_011526774.2 1576 Intron XP_011525076.1
XM_011526775.1 1576 Intron XP_011525077.1
XM_011526776.2 1576 Intron XP_011525078.1
XM_011526777.2 1576 Intron XP_011525079.1
XM_011526778.2 1576 Intron XP_011525080.1
XM_011526779.2 1576 Intron XP_011525081.1
XM_011526782.2 1576 Intron XP_011525084.1
XM_011526783.2 1576 Intron XP_011525085.1
XM_011526784.2 1576 Intron XP_011525086.1
XM_011526785.2 1576 Intron XP_011525087.1
XM_011526786.2 1576 Intron XP_011525088.1
XM_017026620.1 1576 Intron XP_016882109.1
XM_017026621.1 1576 Intron XP_016882110.1
XM_017026622.1 1576 Intron XP_016882111.1
XM_017026623.1 1576 Intron XP_016882112.1
XM_017026624.1 1576 Intron XP_016882113.1
XM_017026625.1 1576 Intron XP_016882114.1
XM_017026626.1 1576 Missense Mutation CGG,TGG R196W XP_016882115.1
XM_017026627.1 1576 Missense Mutation CGG,TGG R162W XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details