Product Details

SNP ID
rs199690352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55159192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGCTCACAGTGTGGGGTCCCA[C/T]CCTGCAGAGGCTGGGTCAGGCCCTC
Phenotype
MIM: 614566 MIM: 191044 MIM: 191041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256714.1 1711 Missense Mutation GAT,GGT D566G NP_001243643.1
NM_001256715.1 1711 Missense Mutation GAT,GGT D499G NP_001243644.1
NM_001256716.1 1711 Missense Mutation GAT,GGT D445G NP_001243645.1
NM_178837.4 1711 Missense Mutation GAT,GGT D546G NP_849159.2
Gene
TNNI3
Gene Name
troponin I3, cardiac type
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
There are no transcripts associated with this gene.

View Full Product Details