Product Details

SNP ID
rs199868688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7608155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCGCAAGGACCGTGTGGTGGC[A/G]CGACGTGCCCCCTGCTTCCCGACGG
Phenotype
MIM: 612685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMSAP3 PubMed Links

Gene Details

Gene
CAMSAP3
Gene Name
calmodulin regulated spectrin associated protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080429.2 806 Silent Mutation GCA,GCG A244A NP_001073898.1
NM_020902.1 806 Silent Mutation GCA,GCG A217A NP_065953.1
XM_005272491.3 806 Silent Mutation GCA,GCG A233A XP_005272548.1
XM_011528154.2 806 Silent Mutation GCA,GCG A244A XP_011526456.1
XM_017027036.1 806 Silent Mutation GCA,GCG A244A XP_016882525.1
Gene
MIR6792
Gene Name
microRNA 6792
There are no transcripts associated with this gene.

View Full Product Details