Product Details

SNP ID
rs199588409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49818465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCCGCAAGCACTACCTGCTC[C/T]CGGCCATCGAGTGAGTGCTGTTTCC
Phenotype
MIM: 610622 MIM: 610197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUZ PubMed Links

Gene Details

Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
There are no transcripts associated with this gene.

Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 187 Missense Mutation CCG,TCG P42S NP_112235.2
XM_011527353.1 187 Missense Mutation CCG,TCG P42S XP_011525655.1

View Full Product Details