Product Details

SNP ID
rs199871271
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7520442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACAAACGGGTGCATGAGGGTGC[A/G]GCCGCTGCTGCAGCTGCCGCGGCCG
Phenotype
MIM: 605248
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MCOLN1 PubMed Links

Gene Details

Gene
MCOLN1
Gene Name
mucolipin 1
There are no transcripts associated with this gene.

Gene
ZNF358
Gene Name
zinc finger protein 358
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018083.4 1261 Silent Mutation GCA,GCG A400A NP_060553.4
XM_005272460.3 1261 Silent Mutation GCA,GCG A400A XP_005272517.1
XM_011527695.1 1261 Silent Mutation GCA,GCG A400A XP_011525997.1
XM_017026296.1 1261 Silent Mutation GCA,GCG A419A XP_016881785.1

View Full Product Details