Product Details

SNP ID
rs200409726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:31276837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGATGTACGTGGAAGGAGTCC[C/T]GATTTGGGACGCACAATCGTTACAA
Phenotype
MIM: 614119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSHZ3 PubMed Links

Gene Details

Gene
TSHZ3
Gene Name
teashirt zinc finger homeobox 3
There are no transcripts associated with this gene.

View Full Product Details