Product Details
- SNP ID
-
rs199837560
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45890916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGAGGTGGAGGCAGGGAGTCAA[C/G]TGGGGTTCCTGGCAGAAGGGGCAGC
- Phenotype
-
MIM: 615331
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IRF2BP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62111265] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IRF2BP1
- Gene Name
- interferon regulatory factor 2 binding protein 1
There are no transcripts associated with this gene.
- Gene
- MYPOP
- Gene Name
- Myb related transcription factor, partner of profilin
View Full Product Details