Product Details

SNP ID
rs199595024
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18386321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGCAAGTTTCCCGGGACCCTC[A/C]GAGTTGCACTCCGAAGACTCCAGAT
Phenotype
MIM: 605312 MIM: 607518
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GDF15 PubMed Links
Additional Information
For this assay, SNP(s) [rs6413435] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GDF15
Gene Name
growth differentiation factor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004864.2 164 Silent Mutation TCA,TCC S44S NP_004855.2
Gene
LRRC25
Gene Name
leucine rich repeat containing 25
There are no transcripts associated with this gene.

Gene
MIR3189
Gene Name
microRNA 3189
There are no transcripts associated with this gene.

View Full Product Details