Product Details

SNP ID
rs200294399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44082072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAGAAGAAAAGTTTTGGATC[A/G]TGGAGACAGCAACCCAAAGAGAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF284 PubMed Links

Gene Details

Gene
ZNF284
Gene Name
zinc finger protein 284
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011526907.2 519 Missense Mutation ATG,GTG M68V XP_011525209.1
XM_011526908.2 519 Missense Mutation ATG,GTG M68V XP_011525210.1

View Full Product Details