Product Details

SNP ID
rs201439283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18784307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCACCTGGGACTCTGTGTCTCC[C/T]GTATGCCACAGAGCGTTCCGCAGCT
Phenotype
MIM: 600310 MIM: 607536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COMP PubMed Links

Gene Details

Gene
COMP
Gene Name
cartilage oligomeric matrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000095.2 2007 Silent Mutation ACA,ACG T657T NP_000086.2
Gene
CRTC1
Gene Name
CREB regulated transcription coactivator 1
There are no transcripts associated with this gene.

View Full Product Details