Product Details

SNP ID
rs201058848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34740783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGAAGGGAAGCATGGAAGTCA[A/G]GTAGACCATTTCAGACCAGCAATTC
Phenotype
MIM: 606741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF181 PubMed Links

Gene Details

Gene
ZNF181
Gene Name
zinc finger protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029997.3 633 Silent Mutation CAA,CAG Q134Q NP_001025168.2
NM_001145665.1 633 Silent Mutation CAA,CAG Q133Q NP_001139137.1
XM_005258849.2 633 Silent Mutation CAA,CAG Q134Q XP_005258906.1
XM_005258850.2 633 Silent Mutation CAA,CAG Q133Q XP_005258907.1
XM_005258851.3 633 Silent Mutation CAA,CAG Q130Q XP_005258908.1
XM_006723183.3 633 Silent Mutation CAA,CAG Q134Q XP_006723246.1
XM_006723184.3 633 Silent Mutation CAA,CAG Q129Q XP_006723247.1
XM_017026739.1 633 Silent Mutation CAA,CAG Q133Q XP_016882228.1
XM_017026740.1 633 Silent Mutation CAA,CAG Q148Q XP_016882229.1

View Full Product Details