Product Details
- SNP ID
-
rs201230520
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4249291 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACTCAAGCTCCCCAAAGACCGG[C/T]AGTACGTGGTGCGGCTGATGGCCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC94
PubMed Links
Gene Details
- Gene
- CCDC94
- Gene Name
- coiled-coil domain containing 94
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018074.4 |
121 |
Nonsense Mutation |
CAG,TAG |
Q30* |
NP_060544.2 |
View Full Product Details