Product Details

SNP ID
rs201891170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10089914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCACAGGCAGTGGCGACCTCCC[C/T]CCTGCCACTGACAGAAGCCAACCTA
Phenotype
MIM: 609336 MIM: 616808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL6 PubMed Links

Gene Details

Gene
ANGPTL6
Gene Name
angiopoietin like 6
There are no transcripts associated with this gene.

Gene
C19orf66
Gene Name
chromosome 19 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308277.1 560 Missense Mutation CCC,CTC P84L NP_001295206.1
NM_018381.3 560 Missense Mutation CCC,CTC P84L NP_060851.2
XM_011528121.1 560 Missense Mutation CCC,CTC P45L XP_011526423.1
XM_011528122.1 560 Intron XP_011526424.1
XM_017026934.1 560 Missense Mutation CCC,CTC P33L XP_016882423.1

View Full Product Details