Product Details

SNP ID
rs200891222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGCCTTTCCCACACACGTTG[C/T]ATCGAAATGGTTTCTCTCCTGTATG
Phenotype
MIM: 604749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 5709 Missense Mutation TAC,TGC Y517C NP_004225.3
XM_005259412.4 5709 Missense Mutation TAC,TGC Y513C XP_005259469.1

View Full Product Details