Product Details

SNP ID
rs201443263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48664736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAACAGCTCAGAGTTGAACCG[C/G]CAGCGTCGGGCGTGCTGGTTGCAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NTN5 PubMed Links

Gene Details

Gene
NTN5
Gene Name
netrin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145807.2 871 Missense Mutation TGC,TGG C221W NP_665806.1
XM_011526443.2 871 Missense Mutation TGC,TGG C221W XP_011524745.1
XM_017026274.1 871 Missense Mutation TGC,TGG C66W XP_016881763.1
Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details