Product Details

SNP ID
rs202103984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43362250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGTGGGGCTGAGGGCCTGGAGT[C/G]TCTCACTTGGGGGGTGGGGCTGGCA
Phenotype
MIM: 162860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD177 PubMed Links

Gene Details

Gene
CD177
Gene Name
CD177 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020406.3 1300 Missense Mutation TCT,TGT S415C NP_065139.2
XM_017027021.1 1300 Intron XP_016882510.1
XM_017027022.1 1300 Intron XP_016882511.1

View Full Product Details