Product Details
- SNP ID
-
rs200858220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1417515 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATCGCAGGAAGCTCTTCGTGGG[C/T]GGTCTTGACTGGAGCACGACCCAAG
- Phenotype
-
MIM: 607430
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DAZAP1
PubMed Links
Gene Details
- Gene
- DAZAP1
- Gene Name
- DAZ associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018959.3 |
203 |
Silent Mutation |
GGC,GGT |
G15G |
NP_061832.2 |
NM_170711.2 |
203 |
Silent Mutation |
GGC,GGT |
G15G |
NP_733829.1 |
XM_005259531.3 |
203 |
Silent Mutation |
GGC,GGT |
G15G |
XP_005259588.1 |
XM_005259534.3 |
203 |
UTR 5 |
|
|
XP_005259591.1 |
XM_005259535.3 |
203 |
UTR 5 |
|
|
XP_005259592.1 |
XM_005259536.3 |
203 |
UTR 5 |
|
|
XP_005259593.1 |
XM_011527904.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526206.1 |
XM_011527905.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526207.1 |
XM_011527906.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526208.1 |
XM_011527907.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526209.1 |
XM_011527908.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526210.1 |
XM_011527909.2 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_011526211.1 |
XM_011527910.2 |
203 |
UTR 5 |
|
|
XP_011526212.1 |
XM_017026581.1 |
203 |
Silent Mutation |
GGC,GGT |
G15G |
XP_016882070.1 |
XM_017026582.1 |
203 |
Silent Mutation |
GGC,GGT |
G158G |
XP_016882071.1 |
View Full Product Details