Product Details

SNP ID
rs201271072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57634650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGGAAGTGTGACCTTTGAAGAT[A/G]TGGCCGTGTACTTCTCCTGGGAGGA
Phenotype
MIM: 604076 MIM: 601856
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
There are no transcripts associated with this gene.

Gene
ZNF211
Gene Name
zinc finger protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265597.2 331 Missense Mutation ATG,GTG M51V NP_001252526.1
NM_001265598.2 331 Missense Mutation ATG,GTG M38V NP_001252527.1
NM_001265599.2 331 UTR 5 NP_001252528.1
NM_001265600.2 331 UTR 5 NP_001252529.1
NM_001322306.1 331 Intron NP_001309235.1
NM_006385.4 331 Missense Mutation ATG,GTG M51V NP_006376.2
NM_198855.3 331 Missense Mutation ATG,GTG M38V NP_942152.1

View Full Product Details