Product Details

SNP ID
rs200516938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49828483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGTGGTGAGAGCTGCAGCCTC[A/G]TCGCGGAAGGACTCAGCACAGCCTT
Phenotype
MIM: 610197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED25 PubMed Links

Gene Details

Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 403 Missense Mutation ATC,GTC I114V NP_112235.2
XM_011527353.1 403 Missense Mutation ATC,GTC I114V XP_011525655.1
Gene
MIR6800
Gene Name
microRNA 6800
There are no transcripts associated with this gene.

View Full Product Details