Product Details
- SNP ID
-
rs201757601
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:496530 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCGGACTGGCCCTCCTGCTGGCG[A/G]GGCTTCTGGGGCTCCTCCTCGGTGA
- Phenotype
-
MIM: 102670
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MADCAM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78668504] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MADCAM1
- Gene Name
- mucosal vascular addressin cell adhesion molecule 1
- Gene
- ODF3L2
- Gene Name
- outer dense fiber of sperm tails 3 like 2
There are no transcripts associated with this gene.
View Full Product Details