Product Details

SNP ID
rs201276767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11575119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATCTCCACATAGGCAAAGCCA[A/C]CCAGTGAGTCTTCAGTCCCATAGTG
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 1272 Missense Mutation GGT,GTT G290V NP_001104504.1
NM_001111035.2 1272 Missense Mutation GGT,GTT G290V NP_001104505.1
NM_001111036.2 1272 Missense Mutation GGT,GTT G290V NP_001104506.1
NM_001322023.1 1272 Missense Mutation GGT,GTT G290V NP_001308952.1
NM_001611.4 1272 Missense Mutation GGT,GTT G290V NP_001602.1
XM_005259938.1 1272 Missense Mutation GGT,GTT G290V XP_005259995.1
XM_011528069.2 1272 Missense Mutation GGT,GTT G290V XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 1272 Intron NP_001277012.1
NM_001290084.1 1272 Intron NP_001277013.1
NM_001290085.1 1272 Intron NP_001277014.1
NM_145295.3 1272 Intron NP_660338.1
XM_011527780.2 1272 Intron XP_011526082.1

View Full Product Details