Product Details

SNP ID
rs200693684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17816878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTAGGGACAGAGGGTCAGCAG[C/G]TCTTGTTGGGTACAGCCACTGAGGC
Phenotype
MIM: 605863 MIM: 146738 MIM: 600173
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT3 PubMed Links

Gene Details

Gene
B3GNT3
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
There are no transcripts associated with this gene.

Gene
INSL3
Gene Name
insulin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265587.1 440 Missense Mutation ACC,AGC T156S NP_001252516.1
NM_005543.3 440 Missense Mutation GAC,GAG D124E NP_005534.2
Gene
JAK3
Gene Name
Janus kinase 3
There are no transcripts associated with this gene.

View Full Product Details