Product Details

SNP ID
rs200865724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12465139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTGAAAAGTTTGGTAATATCT[A/G]AAAGCTTTTCCACATTGTTTACATT
Phenotype
MIM: 606697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
There are no transcripts associated with this gene.

Gene
ZNF709
Gene Name
zinc finger protein 709
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152601.3 944 Silent Mutation TTC,TTT F261F NP_689814.1

View Full Product Details