Product Details

SNP ID
rs201763027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41847554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCATCAGCCGCTCTCCAACCTG[C/T]GCCCGCTGCCGCAACCATGGTGTCA
Phenotype
MIM: 614806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040283.2 231 Silent Mutation TGC,TGT C42C NP_001035373.1
XM_011527214.1 231 UTR 5 XP_011525516.1
XM_011527215.1 231 UTR 5 XP_011525517.1
XM_011527216.2 231 Silent Mutation TGC,TGT C47C XP_011525518.1
XM_017027123.1 231 Silent Mutation TGC,TGT C47C XP_016882612.1
XM_017027124.1 231 Silent Mutation TGC,TGT C45C XP_016882613.1
XM_017027125.1 231 Silent Mutation TGC,TGT C42C XP_016882614.1
XM_017027126.1 231 Silent Mutation TGC,TGT C42C XP_016882615.1
XM_017027127.1 231 Silent Mutation TGC,TGT C42C XP_016882616.1
XM_017027128.1 231 Silent Mutation TGC,TGT C42C XP_016882617.1
XM_017027129.1 231 Silent Mutation TGC,TGT C47C XP_016882618.1
Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
There are no transcripts associated with this gene.

View Full Product Details