Product Details

SNP ID
rs201832670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54108033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCCGCCTTCCTCACCTGCACC[A/G]GGGCCAGCTCTGGAGTCAGCGCATT
Phenotype
MIM: 603832 MIM: 606862 MIM: 606419 MIM: 609519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
There are no transcripts associated with this gene.

Gene
OSCAR
Gene Name
osteoclast associated, immunoglobulin-like receptor
There are no transcripts associated with this gene.

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
There are no transcripts associated with this gene.

Gene
TFPT
Gene Name
TCF3 fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321792.1 647 Missense Mutation CCG,CTG P203L NP_001308721.1
NM_013342.3 647 Missense Mutation CCG,CTG P212L NP_037474.1
XM_005278261.1 647 Missense Mutation CCG,CTG P92L XP_005278318.1

View Full Product Details