Product Details

SNP ID
rs202152370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6732310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACCTTCTTCTCCTTATCCGACA[A/G]GACGTACTTGATGAAGGCCCAGCCT
Phenotype
MIM: 604504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR108 PubMed Links

Gene Details

Gene
GPR108
Gene Name
G protein-coupled receptor 108
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080452.1 1082 Silent Mutation CTG,TTG L360L NP_001073921.1
XM_006722801.3 1082 Silent Mutation CTG,TTG L118L XP_006722864.1
XM_011528137.2 1082 Silent Mutation CTG,TTG L118L XP_011526439.1
XM_017027012.1 1082 Silent Mutation CTG,TTG L360L XP_016882501.1
XM_017027013.1 1082 Silent Mutation CTG,TTG L360L XP_016882502.1
XM_017027014.1 1082 Silent Mutation CTG,TTG L360L XP_016882503.1
XM_017027015.1 1082 Silent Mutation CTG,TTG L118L XP_016882504.1
Gene
MIR6791
Gene Name
microRNA 6791
There are no transcripts associated with this gene.

Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
There are no transcripts associated with this gene.

View Full Product Details