Product Details

SNP ID
rs200867987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39415160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCGCCTACATCTCTCTCCGGTGG[A/G]GATCCCAGGTTCAGCCAGACCCTCA
Phenotype
MIM: 611893 MIM: 190700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4530 PubMed Links

Gene Details

Gene
MIR4530
Gene Name
microRNA 4530
There are no transcripts associated with this gene.

Gene
PLEKHG2
Gene Name
pleckstrin homology and RhoGEF domain containing G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022835.2 1134 Missense Mutation GAG,GGG E93G NP_073746.2
XM_005259163.2 1134 Missense Mutation GAG,GGG E93G XP_005259220.1
XM_006723334.2 1134 Intron XP_006723397.1
XM_011527232.2 1134 Missense Mutation GAG,GGG E94G XP_011525534.1
XM_017027150.1 1134 Missense Mutation GAG,GGG E94G XP_016882639.1
XM_017027151.1 1134 Missense Mutation GAG,GGG E94G XP_016882640.1
Gene
ZFP36
Gene Name
ZFP36 ring finger protein
There are no transcripts associated with this gene.

View Full Product Details