Product Details

SNP ID
rs201142356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45692788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTCTTGCCGCCGAAGCGCCGC[C/T]TGCTACCGCGGGTTCGGCTCTTGCC
Phenotype
MIM: 137241 MIM: 601061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIPR PubMed Links

Gene Details

Gene
GIPR
Gene Name
gastric inhibitory polypeptide receptor
There are no transcripts associated with this gene.

Gene
QPCTL
Gene Name
glutaminyl-peptide cyclotransferase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163377.1 385 Silent Mutation CTG,TTG L29L NP_001156849.1
NM_017659.3 385 Silent Mutation CTG,TTG L29L NP_060129.2
XM_011527048.2 385 Silent Mutation CTG,TTG L29L XP_011525350.1
XM_017026900.1 385 Silent Mutation CTG,TTG L29L XP_016882389.1
Gene
SNRPD2
Gene Name
small nuclear ribonucleoprotein D2 polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004597.6 385 Intron NP_004588.1
NM_177542.3 385 Intron NP_808210.2

View Full Product Details