Product Details

SNP ID
rs200492063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43626164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTACTGCGTCTGCTTGCTGTGTCC[A/G]GAGGGCAGCGCCTGGTTCTGCGCCT
Phenotype
MIM: 609744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145296.1 582 Silent Mutation TCC,TCT S208S NP_660339.1
XM_005258620.2 582 Silent Mutation TCC,TCT S208S XP_005258677.1
XM_017026452.1 582 Silent Mutation TCC,TCT S189S XP_016881941.1
Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

View Full Product Details