Product Details
- SNP ID
-
rs201354847
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57808488 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTTTTTCTCTAGTGTGAACTCCT[C/T]GATGATTACTGAAGCTGTCATATTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF552
PubMed Links
Gene Details
- Gene
- ZNF552
- Gene Name
- zinc finger protein 552
View Full Product Details