Product Details

SNP ID
rs202013571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13933168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGCAGCACCAGGGCACGCAGG[C/T]GGCGGGGCAGGGCTGGAGGCACGCG
Phenotype
MIM: 610055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CC2D1A PubMed Links

Gene Details

Gene
CC2D1A
Gene Name
coiled-coil and C2 domain containing 1A
There are no transcripts associated with this gene.

Gene
PODNL1
Gene Name
podocan like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146254.1 1184 Missense Mutation CAC,CGC H357R NP_001139726.1
NM_001146255.1 1184 Missense Mutation CAC,CGC H268R NP_001139727.1
NM_024825.3 1184 Missense Mutation CAC,CGC H359R NP_079101.3
XM_006722901.1 1184 Missense Mutation CAC,CGC H359R XP_006722964.1
XM_006722902.1 1184 Missense Mutation CAC,CGC H314R XP_006722965.1
XM_006722903.1 1184 Missense Mutation CAC,CGC H270R XP_006722966.1
XM_011528308.1 1184 Missense Mutation CAC,CGC H357R XP_011526610.1
XM_011528309.2 1184 Missense Mutation CAC,CGC H341R XP_011526611.1
XM_011528310.2 1184 Missense Mutation CAC,CGC H329R XP_011526612.1
XM_011528311.1 1184 Missense Mutation CAC,CGC H326R XP_011526613.1
XM_011528313.1 1184 Missense Mutation CAC,CGC H277R XP_011526615.1
XM_011528314.1 1184 Missense Mutation CAC,CGC H262R XP_011526616.1
XM_011528316.1 1184 Missense Mutation CAC,CGC H188R XP_011526618.1
XM_017027313.1 1184 Missense Mutation CAC,CGC H262R XP_016882802.1

View Full Product Details