Product Details

SNP ID
rs202017019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57492916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGATTGCTTCTGTAGGACTGC[C/T]CAGTTCAAACATTCAGCAACACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF419 PubMed Links

Gene Details

Gene
ZNF419
Gene Name
zinc finger protein 419
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098491.1 563 Missense Mutation CCC,CTC P121L NP_001091961.1
NM_001098492.1 563 Missense Mutation CCC,CTC P108L NP_001091962.1
NM_001098493.1 563 Missense Mutation CCC,CTC P107L NP_001091963.1
NM_001098494.1 563 Missense Mutation CCC,CTC P88L NP_001091964.1
NM_001098495.1 563 Missense Mutation CCC,CTC P75L NP_001091965.1
NM_001098496.1 563 Missense Mutation CCC,CTC P74L NP_001091966.1
NM_001291743.1 563 Missense Mutation CCC,CTC P87L NP_001278672.1
NM_001291744.1 563 Missense Mutation CCC,CTC P32L NP_001278673.1
NM_001291745.1 563 Intron NP_001278674.1
NM_024691.3 563 Missense Mutation CCC,CTC P120L NP_078967.3
Gene
ZNF773
Gene Name
zinc finger protein 773
There are no transcripts associated with this gene.

View Full Product Details