Product Details
- SNP ID
-
rs201951170
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44172110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGAAGCAGTGACCTTCAAGGACG[C/T]GGCTGTGGCCTTCACGGAGGAGGAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF226
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2232801] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF226
- Gene Name
- zinc finger protein 226
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001032372.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001027544.1 |
NM_001032373.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001027545.1 |
NM_001032374.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001027546.1 |
NM_001146220.2 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001139692.1 |
NM_001319088.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001306017.1 |
NM_001319089.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001306018.1 |
NM_001319090.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_001306019.1 |
NM_015919.3 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_057003.2 |
NM_016444.2 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
NP_057528.2 |
XM_005259227.2 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_005259284.1 |
XM_006723367.3 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_006723430.1 |
XM_006723368.3 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_006723431.1 |
XM_006723369.2 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_006723432.1 |
XM_017027262.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_016882751.1 |
XM_017027263.1 |
343 |
UTR 5 |
|
|
XP_016882752.1 |
XM_017027264.1 |
343 |
UTR 5 |
|
|
XP_016882753.1 |
XM_017027265.1 |
343 |
Missense Mutation |
GCG,GTG |
A13V |
XP_016882754.1 |
View Full Product Details