Product Details

SNP ID
rs202229314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56182177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCCCTCCCTACCCAGTCCTC[C/T]ACCTTCCCCAAATGGGTGACCAAGA
Phenotype
MIM: 611178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145546.1 224 Silent Mutation TCC,TCT S31S NP_001139018.1
NM_033106.3 224 Missense Mutation CAC,TAC H48Y NP_149097.1
Gene
ZSCAN5B
Gene Name
zinc finger and SCAN domain containing 5B
There are no transcripts associated with this gene.

View Full Product Details