Product Details

SNP ID
rs200673226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55159099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAGTCTGACTCACAGTTGGGC[A/G]GAGCCAAGGCCCCCTGAGGCTGAGC
Phenotype
MIM: 614566 MIM: 191044 MIM: 191041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256714.1 1804 Missense Mutation CCG,CTG P597L NP_001243643.1
NM_001256715.1 1804 Missense Mutation CCG,CTG P530L NP_001243644.1
NM_001256716.1 1804 Missense Mutation CCG,CTG P476L NP_001243645.1
NM_178837.4 1804 Missense Mutation CCG,CTG P577L NP_849159.2
Gene
TNNI3
Gene Name
troponin I3, cardiac type
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
There are no transcripts associated with this gene.

View Full Product Details