Product Details

SNP ID
rs200709260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41264693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACCCGGGCACATCAACGAGGAG[G/T]TCGAGACCGAGGGGGGCTCCGAGCT
Phenotype
MIM: 109135 MIM: 605800
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
HNRNPUL1
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301016.2 520 Intron NP_001287945.1
NM_001321208.1 520 Intron NP_001308137.1
NM_001321211.1 520 Intron NP_001308140.1
NM_007040.5 520 Missense Mutation GTC,TTC V64F NP_008971.2
NM_144732.4 520 Intron NP_653333.1
XM_005258459.3 520 Missense Mutation GTC,TTC V64F XP_005258516.1
XM_005258461.4 520 Intron XP_005258518.1
XM_005258463.3 520 Intron XP_005258520.1
XM_005258464.3 520 Intron XP_005258521.1
XM_005258465.4 520 Intron XP_005258522.1
XM_011526392.2 520 Missense Mutation GTC,TTC V64F XP_011524694.1
XM_011526393.2 520 Intron XP_011524695.1
XM_011526394.2 520 Intron XP_011524696.1
XM_011526395.2 520 Intron XP_011524697.1

View Full Product Details