Product Details
- SNP ID
-
rs201188525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11777737 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCATCAGAAACCTGGACTGTATA[A/G]GTAAGGATGTCATCATGTCTTCACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF441
PubMed Links
Gene Details
- Gene
- ZNF441
- Gene Name
- zinc finger protein 441
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152355.2 |
324 |
Missense Mutation |
AGA,GGA |
R44G |
NP_689568.2 |
View Full Product Details