Product Details

SNP ID
rs201638495
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:226152345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAAGGGTTCTACCTTCTTCCTC[C/G]TCGTCGTCATCGCTGGACTCACTGA
Phenotype
MIM: 606809
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACBD3 PubMed Links

Gene Details

Gene
ACBD3
Gene Name
acyl-CoA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022735.3 1412 Missense Mutation GAC,GAG D455E NP_073572.2

View Full Product Details