Product Details

SNP ID
rs201282280
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:16447892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGAGCTCTTTGCTCAGGCCCC[A/G]GTGGATCAGTTTCCTGGCACAGCTG
Phenotype
MIM: 611624
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECAP2 PubMed Links

Gene Details

Gene
NECAP2
Gene Name
NECAP endocytosis associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145277.1 306 Silent Mutation CCA,CCG P72P NP_001138749.1
NM_001145278.1 306 Silent Mutation CCA,CCG P46P NP_001138750.1
NM_018090.4 306 Silent Mutation CCA,CCG P72P NP_060560.1
XM_011541767.2 306 Silent Mutation CCA,CCG P19P XP_011540069.1
XM_017001721.1 306 Silent Mutation CCA,CCG P72P XP_016857210.1

View Full Product Details