Product Details

SNP ID
rs201296195
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:203483379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCCTCCTCCCCTCCCTCCAGG[C/T]CCTCCATCTATCTTCCCTGACTGTC
Phenotype
MIM: 601914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRELP PubMed Links

Gene Details

Gene
PRELP
Gene Name
proline and arginine rich end leucine rich repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002725.3 395 Silent Mutation GGC,GGT G65G NP_002716.1
NM_201348.1 395 Silent Mutation GGC,GGT G65G NP_958505.1

View Full Product Details