Product Details

SNP ID
rs201297278
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:43655755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCGAGAGTTCCGCAAGATAGC[C/T]AATAGCGATAAGTGAGTGGAAACCC
Phenotype
MIM: 609764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 506 Silent Mutation GCC,GCT A101A NP_055478.2
XM_005271354.3 506 Silent Mutation GCC,GCT A101A XP_005271411.1
XM_005271355.3 506 Silent Mutation GCC,GCT A101A XP_005271412.1
XM_005271356.3 506 Silent Mutation GCC,GCT A10A XP_005271413.1
XM_011542467.2 506 Intron XP_011540769.2
XM_017002912.1 506 Silent Mutation GCC,GCT A10A XP_016858401.1

View Full Product Details