Product Details

SNP ID
rs201231728
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156009545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCTGTGGAAGCCCCAATCAGT[C/T]CTTCTTCGTTTTGGGAGTGTCATAT
Phenotype
MIM: 600867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSR2 PubMed Links

Gene Details

Gene
SSR2
Gene Name
signal sequence receptor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003145.3 627 Missense Mutation AAC,GAC N183D NP_003136.1
XM_011509923.2 627 Missense Mutation AAC,GAC N202D XP_011508225.1

View Full Product Details