Product Details

SNP ID
rs201297417
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:31630836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCAGGTTGTAGCCCATTTGG[A/G]ACAGAGCTGGAGAAGAGGGGCACAC
Phenotype
MIM: 602392 MIM: 610033
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR1 PubMed Links

Gene Details

Gene
HCRTR1
Gene Name
hypocretin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001525.2 974 Intron NP_001516.2
XM_017001105.1 974 Intron XP_016856594.1
XM_017001106.1 974 Intron XP_016856595.1
XM_017001107.1 974 Intron XP_016856596.1
Gene
PEF1
Gene Name
penta-EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012392.3 974 Missense Mutation TCC,TTC S211F NP_036524.1
XM_011541745.1 974 Missense Mutation TCC,TTC S206F XP_011540047.1
XM_011541746.2 974 Missense Mutation TCC,TTC S141F XP_011540048.1
XM_011541747.1 974 Missense Mutation TCC,TTC S141F XP_011540049.1
XM_017001680.1 974 Missense Mutation TCC,TTC S141F XP_016857169.1
XM_017001681.1 974 Intron XP_016857170.1

View Full Product Details